Gene Transcripts

Gene Symbol
Wbscr25
Entrez Gene ID
71304
Gene Name
Williams Beuren syndrome chromosome region 25 (human)
Gene Aliases
4933433J03Rik, 4933439O14Rik
Chromosome Location
Chr. 5: 134987433 - 135001350 on Build GRCm38
UniGene ID
Mm.482317
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026907.1 1 1209
GenBank mRNA AK017044.1 - 1 821
AK017130.1 - 1 368
AY081954.2 - 1 1209

View Full Product Details