Gene Transcripts
- Gene Symbol
- Wbscr25
- Entrez Gene ID
- 71304
- Gene Name
- Williams Beuren syndrome chromosome region 25 (human)
- Gene Aliases
- 4933433J03Rik, 4933439O14Rik
- Chromosome Location
- Chr. 5: 134987433 - 135001350 on Build GRCm38
- UniGene ID
- Mm.482317
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026907.1 |
|
1 |
710 |
|
GenBank mRNA |
AK017044.1 |
- |
1 |
323 |
|
AY081954.2 |
- |
1 |
710 |
|
View Full Product Details