Gene Transcripts
- Gene Symbol
- Gm5
- Entrez Gene ID
- 194292
- Gene Name
- predicted gene 5
- Gene Aliases
- Gm1812
- Chromosome Location
- Chr. 5: 150499747 - 150502219 on Build GRCm38
- UniGene ID
- Mm.300184
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024513.1 |
|
1 |
88 |
|
GenBank mRNA |
CO797249.1 |
- |
1 |
117 |
|
View Full Product Details