Gene Transcripts
- Gene Symbol
- NHEG1
- Entrez Gene ID
- 100294720
- Gene Name
- neuroblastoma highly expressed 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 136982159 - 136993231 on Build GRCh38
- UniGene ID
- Hs.520412
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AL522654.3 |
- |
3 |
577 |
|
Chr. 6 - 136982640 - 136993231 [-] |
GenBankmRNA |
BF206844.1 |
- |
|
576 |
|
Chr. - - [-] |
View Full Product Details