Gene Transcripts
- Gene Symbol
- HIGD2B
- Entrez Gene ID
- 123346
- Gene Name
- HIG1 hypoxia inducible domain family member 2B
- Gene Aliases
- HIGD2BP
- Chromosome Location
- Chr. 15: 72675782 - 72686149 on Build GRCh38
- UniGene ID
- Hs.434111
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002780.1 |
|
1 |
87 |
|
GenBank mRNA |
BC040890.1 |
- |
1 |
87 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM564128.1 |
- |
1 |
107 |
|
Chr. 15 - 72675889 - 72686150 [-] |
View Full Product Details