Gene Transcripts
- Gene Symbol
- JMJD7-PLA2G4B
- Entrez Gene ID
- 8681
- Gene Name
- JMJD7-PLA2G4B readthrough
- Gene Aliases
- HsT16992, cPLA2-beta
- Chromosome Location
- Chr. 15: 41828085 - 41848148 on Build GRCh38
- UniGene ID
- Hs.198161
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209150.1 |
- |
7 |
3386 |
|
AF065216.1 |
- |
7 |
3223 |
|
BC128426.1 |
- |
1 |
242 |
|
View Full Product Details