Gene Transcripts
- Gene Symbol
- BACH1
- Entrez Gene ID
- 571
- Gene Name
- BTB domain and CNC homolog 1
- Gene Aliases
- BACH-1, BTBD24
- Chromosome Location
- Chr. 21: 29298795 - 29361896 on Build GRCh38
- UniGene ID
- Hs.154276
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027655.2 |
|
7 |
2298 |
|
GenBank mRNA |
AF317902.1 |
- |
3 |
343 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000504298.1 |
- |
3 |
343 |
|
Chr. 21 - 29351634 - 29361894 [+] |
View Full Product Details