Gene Transcripts
- Gene Symbol
- ARMCX4
- Entrez Gene ID
- 100131755
- Gene Name
- armadillo repeat containing, X-linked 4
- Gene Aliases
- CXorf35, GASP4
- Chromosome Location
- Chr. X: 101418263 - 101535988 on Build GRCh38
- UniGene ID
- Hs.729062
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028407.2 |
|
16 |
4110 |
|
GenBank mRNA |
CR933662.1 |
- |
16 |
4085 |
|
View Full Product Details