Gene Transcripts

Gene Symbol
EFHC1
Entrez Gene ID
114327
Gene Name
EF-hand domain containing 1
Gene Aliases
EJM1, dJ304B14.2
Chromosome Location
Chr. 6: 52420196 - 52495785 on Build GRCh38
UniGene ID
Hs.403171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033327.1 4 1287
GenBank mRNA AK225543.1 - 4 1181
AL122084.1 - 4 1174
AY608690.1 - 4 1741

View Full Product Details