Gene Transcripts
- Gene Symbol
- GSTTP1
- Entrez Gene ID
- 25774
- Gene Name
- glutathione S-transferase theta pseudogene 1
- Gene Aliases
- GSTT4, HS322B1A
- Chromosome Location
- Chr. 22: 23998401 - 24005361 on Build GRCh38
- UniGene ID
- Hs.652490
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY005337.1 |
- |
1, 2 |
195 |
|
Chr. 22|NT_187633.1 - 236472 - 241509 [-] |
GenBankmRNA |
HY028266.1 |
- |
1, 2 |
195 |
|
Chr. 22|NT_187633.1 - 235033 - 241509 [-] |
GenBankmRNA |
HY019480.1 |
- |
1, 2 |
225 |
|
Chr. 22|NT_187633.1 - 236451 - 241538 [-] |
View Full Product Details