Gene Transcripts
- Gene Symbol
- ANAPC13
- Entrez Gene ID
- 25847
- Gene Name
- anaphase promoting complex subunit 13
- Gene Aliases
- APC13, SWM1
- Chromosome Location
- Chr. 3: 134477704 - 134486023 on Build GRCh38
- UniGene ID
- Hs.106909
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM459488.1 |
- |
|
107 |
|
Chr. - - [-] |
GenBankmRNA |
D52765.1 |
- |
1 |
143 |
|
Chr. 3 - 134478632 - 134485954 [-] |
GenBankmRNA |
D55213.1 |
- |
1 |
143 |
|
Chr. 3 - 134478681 - 134485954 [-] |
GenBankmRNA |
HY181801.1 |
- |
1 |
175 |
|
Chr. 3 - 134478653 - 134485985 [-] |
GenBankmRNA |
AL550326.3 |
- |
1 |
161 |
|
Chr. 3 - 134478584 - 134485972 [-] |
GenBankmRNA |
CB994657.1 |
- |
|
200 |
|
Chr. - - [-] |
GenBankmRNA |
BM807200.1 |
- |
1 |
171 |
|
Chr. 3 - 134478128 - 134485982 [-] |
View Full Product Details