Gene Transcripts

Gene Symbol
PART1
Entrez Gene ID
25859
Gene Name
prostate androgen-regulated transcript 1 (non-protein coding)
Gene Aliases
NCRNA00206
Chromosome Location
Chr. 5: 60487713 - 60547657 on Build GRCh38
UniGene ID
Hs.146312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024617.1 1 274
NR_028508.1 1 55
NR_028509.1 1 55
GenBank mRNA AF163475.1 - 1 55
AK312071.1 - 1 277

View Full Product Details