Gene Transcripts

Gene Symbol
ADAM3A
Entrez Gene ID
1587
Gene Name
ADAM metallopeptidase domain 3A (pseudogene)
Gene Aliases
ADAM3, CYRN1, tMDCI
Chromosome Location
Chr. 8: 39451045 - 39522989 on Build GRCh38
UniGene ID
Hs.98848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001569.3 11 1260
NR_024106.2 11 1260
NR_024107.2 11 1260
NR_073423.1 11 1260
GenBank mRNA AK302269.1 - 8 807
AK308250.1 - 11 1124


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI829329.1 - 7 688 Chr. 8 - 39490262 - 39522812 [-]
GenBankmRNA BM908226.1 - 9 862 Chr. 8 - 39475427 - 39522839 [-]

View Full Product Details