Gene Transcripts
- Gene Symbol
- TSEN15
- Entrez Gene ID
- 116461
- Gene Name
- tRNA splicing endonuclease subunit 15
- Gene Aliases
- C1orf19, PCH2F, sen15
- Chromosome Location
- Chr. 1: 184050207 - 184095846 on Build GRCh38
- UniGene ID
- Hs.548197
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023349.2 |
|
4, 5 |
569 |
|
GenBank mRNA |
AK307241.1 |
- |
4, 5 |
460 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU166129.1 |
- |
3, 4 |
289 |
|
Chr. 1 - 184054394 - 184073115 [+] |
GenBankmRNA |
DA666304.1 |
- |
4, 5 |
460 |
|
Chr. 1 - 184051760 - 184072236 [+] |
View Full Product Details