Gene Transcripts
- Gene Symbol
- SNORA11C
- Entrez Gene ID
- 100124540
- Gene Name
- small nucleolar RNA, H/ACA box 11C
- Gene Aliases
- Chromosome Location
- Chr. X: 47388650 - 47388776 on Build GRCh38
- UniGene ID
- Hs.693443
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003710.1 |
|
1 |
40 |
|
View Full Product Details