Gene Transcripts
- Gene Symbol
- TTTY12
- Entrez Gene ID
- 83867
- Gene Name
- testis-specific transcript, Y-linked 12 (non-protein coding)
- Gene Aliases
- NCRNA00135, TTY11, TTY12
- Chromosome Location
- Chr. Y: 7804924 - 7810683 on Build GRCh38
- UniGene ID
- Hs.667616
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001551.1 |
|
2 |
520 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF332241.3 |
- |
2 |
520 |
|
Chr. Y - 7804924 - 7810683 [+] |
View Full Product Details