Gene Transcripts
- Gene Symbol
- FAM95B1
- Entrez Gene ID
- 100133036
- Gene Name
- family with sequence similarity 95 member B1
- Gene Aliases
- Chromosome Location
- Chr. 9: 40323571 - 40329220 on Build GRCh38
- UniGene ID
- Hs.584289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026759.1 |
|
4 |
1069 |
|
GenBank mRNA |
AK125850.1 |
- |
2 |
3604 |
|
AL833349.1 |
- |
1 |
1030 |
|
- Gene Symbol
- ANKRD18A
- Entrez Gene ID
- 253650
- Gene Name
- ankyrin repeat domain 18A
- Gene Aliases
- Chromosome Location
- Chr. 9: 38566260 - 38620587 on Build GRCh38
- UniGene ID
- Hs.644630
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093059.1 |
- |
7 |
1260 |
|
BC131497.1 |
- |
7 |
1357 |
|
- Gene Symbol
- FAM95A
- Entrez Gene ID
- 90499
- Gene Name
- family with sequence similarity 95 member A
- Gene Aliases
- Chromosome Location
- Chr. 2: 94755326 - 94761184 on Build GRCh38
- UniGene ID
- Hs.458439
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038409.1 |
|
1 |
1030 |
|
GenBank mRNA |
AL137712.1 |
- |
1 |
1030 |
|
- Gene Symbol
- FAM95C
- Entrez Gene ID
- 100289137
- Gene Name
- family with sequence similarity 95 member C
- Gene Aliases
- Chromosome Location
- Chr. 9: 38540567 - 38545372 on Build GRCh38
- UniGene ID
- Hs.644630
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_047651.1 |
|
2 |
2217 |
|
GenBank mRNA |
AK097951.1 |
- |
2 |
2219 |
|
- Gene Symbol
- LOC105379252
- Entrez Gene ID
- 105379252
- Gene Name
- family with sequence similarity 95 member B1-like
- Gene Aliases
- Chromosome Location
- Chr. 9: 66047085 - 66052734 on Build GRCh38
- UniGene ID
- Hs.584289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136310.1 |
|
4 |
1069 |
|
View Full Product Details