Gene Transcripts

Gene Symbol
BBIP1
Entrez Gene ID
92482
Gene Name
BBSome interacting protein 1
Gene Aliases
BBIP10, BBS18, NCRNA00081, bA348N5.3
Chromosome Location
Chr. 10: 110898730 - 110919366 on Build GRCh38
UniGene ID
Hs.232543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195304.1 NP_001182233.1 5 1172
NM_001195305.1 NP_001182234.1 4 1015
NM_001195306.1 NP_001182235.1 4 861
NM_001195307.1 NP_001182236.1 3 940
NM_001243783.1 NP_001230712.1 3 922
GenBank mRNA AK025724.1 - 4 835
AK307126.1 - 4 861
AK307314.1 - 5 1048
BC105804.1 - 5 1011


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CX872108.1 - 3 774 Chr. 10 - 110899848 - 110918213 [-]
GenBankmRNA BG204749.1 - 2 681 Chr. 10 - 110899891 - 110901612 [-]
GenBankmRNA HY274961.1 - 1 110 Chr. 10 - 110899553 - 110900013 [-]
GenBankmRNA BX441766.2 - 4 805 Chr. 10 - 110899867 - 110919156 [-]

View Full Product Details