Gene Transcripts

Gene Symbol
SARNP
Entrez Gene ID
84324
Gene Name
SAP domain containing ribonucleoprotein
Gene Aliases
CIP29, HCC1, HSPC316, THO1
Chromosome Location
Chr. 12: 55752463 - 55817756 on Build GRCh38
UniGene ID
Hs.505676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026722.1 11 1134
NR_026723.1 12 1181
GenBank mRNA AK290508.1 - 12 1149

View Full Product Details