Gene Transcripts
- Gene Symbol
- TCL6
- Entrez Gene ID
- 27004
- Gene Name
- T-cell leukemia/lymphoma 6 (non-protein coding)
- Gene Aliases
- TNG1, TNG2
- Chromosome Location
- Chr. 14: 95651178 - 95673452 on Build GRCh38
- UniGene ID
- Hs.510368
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028288.1 |
|
8 |
2789 |
|
GenBank mRNA |
AB035335.1 |
- |
5 |
3146 |
|
AB035337.1 |
- |
8 |
2792 |
|
View Full Product Details