Gene Transcripts

Gene Symbol
MTX2
Entrez Gene ID
10651
Gene Name
metaxin 2
Gene Aliases
Chromosome Location
Chr. 2: 176269395 - 176338025 on Build GRCh38
UniGene ID
Hs.470728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006635.2 NP_001006636.1 2 423
GenBank mRNA AK303564.1 - 2 348
AK315918.1 - 2 359
BC017271.2 - 2 353 4635884
BG699784.1 - 2 423

View Full Product Details