Gene Transcripts

Gene Symbol
HLA-DPB2
Entrez Gene ID
3116
Gene Name
major histocompatibility complex, class II, DP beta 2 (pseudogene)
Gene Aliases
DP2B, DPB2, DPbeta2, HLA-DP2B
Chromosome Location
Chr. 6: 33112516 - 33129113 on Build GRCh38
UniGene ID
Hs.665450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001435.1 1, 2 109
GenBank mRNA AK301188.1 - 1, 2 148

View Full Product Details