Gene Transcripts

Gene Symbol
WHAMMP3
Entrez Gene ID
339005
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules pseudogene 3
Gene Aliases
WHAMML1, WHDC1L1
Chromosome Location
Chr. 15: 22664739 - 22685368 on Build GRCh38
UniGene ID
Hs.212670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003521.1 7 1496
GenBank mRNA BC048987.1 - 7 1556
DQ309040.1 - 7 1496

Gene Symbol
WHAMMP1
Entrez Gene ID
100288615
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules pseudogene 1
Gene Aliases
WHDC1P1
Chromosome Location
Chr. 15: 32519848 - 32533741 on Build GRCh38
UniGene ID
Hs.562620
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC117556.1 - 1120

View Full Product Details