Gene Transcripts

Gene Symbol
UBE2Q2P1
Entrez Gene ID
388165
Gene Name
ubiquitin conjugating enzyme E2 Q2 pseudogene 1
Gene Aliases
UBE2QP1
Chromosome Location
Chr. 15: 84527196 - 84570795 on Build GRCh38
UniGene ID
Hs.498348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003661.2 6 1787
GenBank mRNA AK125266.1 - 6 1787

Gene Symbol
GOLGA6L9
Entrez Gene ID
440295
Gene Name
golgin A6 family-like 9
Gene Aliases
GOLGA6L20
Chromosome Location
Chr. 15: 82429363 - 82439387 on Build GRCh38
UniGene ID
Hs.498322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK092106.1 - 1 1630

View Full Product Details