Gene Transcripts
- Gene Symbol
- NKAPP1
- Entrez Gene ID
- 158801
- Gene Name
- NFKB activating protein pseudogene 1
- Gene Aliases
- CXorf42
- Chromosome Location
- Chr. X: 120236452 - 120245267 on Build GRCh38
- UniGene ID
- Hs.442518
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG527810.1 |
- |
2 |
458 |
|
Chr. X - 120238193 - 120245207 [-] |
GenBankmRNA |
DA017116.1 |
- |
2 |
462 |
|
Chr. X - 120240280 - 120245211 [-] |
GenBankmRNA |
DB202379.1 |
- |
2 |
483 |
|
Chr. X - 120240252 - 120245233 [-] |
GenBankmRNA |
DA226807.1 |
- |
2 |
461 |
|
Chr. X - 120240244 - 120245211 [-] |
GenBankmRNA |
DA017113.1 |
- |
2 |
461 |
|
Chr. X - 120240281 - 120245211 [-] |
View Full Product Details