Gene Transcripts

Gene Symbol
HAVCR1P1
Entrez Gene ID
100101266
Gene Name
hepatitis A virus cellular receptor 1 pseudogene 1
Gene Aliases
Chromosome Location
Chr. 19: 24162193 - 24163447 on Build GRCh38
UniGene ID
Hs.149312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003603.1 1 1078

Gene Symbol
HAVCR1
Entrez Gene ID
26762
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1
Chromosome Location
Chr. 5: 156979443 - 157059119 on Build GRCh38
UniGene ID
Hs.129711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF043724.1 - 9 1167

View Full Product Details