Gene Transcripts

Gene Symbol
AATK-AS1
Entrez Gene ID
388428
Gene Name
AATK antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 17: 81165507 - 81183164 on Build GRCh38
UniGene ID
Hs.591202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027255.1 3 611
GenBank mRNA AK126811.1 - 3 611
BC140737.1 - 3 602
BC171777.1 - 3 602

View Full Product Details