Gene Transcripts
- Gene Symbol
- PRG1
- Entrez Gene ID
- 23574
- Gene Name
- p53-responsive gene 1
- Gene Aliases
- Chromosome Location
- Chr. 19: 43349056 - 43349548 on Build GRCh38
- UniGene ID
- Hs.721578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026881.1 |
|
1 |
236 |
|
View Full Product Details