Gene Transcripts
- Gene Symbol
- SNORA2B
- Entrez Gene ID
- 677794
- Gene Name
- small nucleolar RNA, H/ACA box 2B
- Gene Aliases
- ACA2b
- Chromosome Location
- Chr. 12: 48667457 - 48667593 on Build GRCh38
- UniGene ID
- Hs.683444
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002951.1 |
|
1 |
17 |
|
GenBank mRNA |
AJ609427.1 |
- |
1 |
17 |
|
View Full Product Details