Gene Transcripts
- Gene Symbol
- ECEL1P2
- Entrez Gene ID
- 347694
- Gene Name
- endothelin converting enzyme like 1 pseudogene 2
- Gene Aliases
- ECEL2
- Chromosome Location
- Chr. 2: 232385750 - 232387044 on Build GRCh38
- UniGene ID
- Hs.452575
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028501.1 |
|
1 |
911 |
|
View Full Product Details