Gene Transcripts
- Gene Symbol
- ANKRD26P3
- Entrez Gene ID
- 100101938
- Gene Name
- ankyrin repeat domain 26 pseudogene 3
- Gene Aliases
- LINC00414
- Chromosome Location
- Chr. 13: 19262800 - 19344973 on Build GRCh38
- UniGene ID
- Hs.129283
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027248.3 |
|
14 |
2143 |
|
GenBank mRNA |
BC036210.1 |
- |
14 |
2143 |
|
View Full Product Details