Gene Transcripts

Gene Symbol
ANKRD26P3
Entrez Gene ID
100101938
Gene Name
ankyrin repeat domain 26 pseudogene 3
Gene Aliases
LINC00414
Chromosome Location
Chr. 13: 19262800 - 19344973 on Build GRCh38
UniGene ID
Hs.129283
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027248.3 14 2427
GenBank mRNA BC036210.1 - 14 2427

View Full Product Details