Gene Transcripts
- Gene Symbol
- SERHL
- Entrez Gene ID
- 94009
- Gene Name
- serine hydrolase-like (pseudogene)
- Gene Aliases
- BK126B4.1, BK126B4.2, HS126B42, dJ222E13.1
- Chromosome Location
- Chr. 22: 42500579 - 42512560 on Build GRCh38
- UniGene ID
- Hs.360940
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027786.1 |
|
9, 10 |
810 |
|
GenBank mRNA |
AK094872.1 |
- |
8, 9 |
1166 |
|
AL365513.1 |
- |
9, 10 |
799 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000455578.5 |
- |
8, 9 |
1166 |
|
Chr. 22 - 42500579 - 42512559 [+] |
View Full Product Details