Gene Transcripts
- Gene Symbol
- SNORA16A
- Entrez Gene ID
- 692073
- Gene Name
- small nucleolar RNA, H/ACA box 16A
- Gene Aliases
- ACA16
- Chromosome Location
- Chr. 1: 28580920 - 28581053 on Build GRCh38
- UniGene ID
- Hs.632377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003035.1 |
|
1 |
93 |
|
GenBank mRNA |
AJ609433.1 |
- |
1 |
93 |
|
- Gene Symbol
- SNHG12
- Entrez Gene ID
- 85028
- Gene Name
- small nucleolar RNA host gene 12
- Gene Aliases
- ASLNC04080, C1orf79, LINC00100, NCRNA00100
- Chromosome Location
- Chr. 1: 28578538 - 28581854 on Build GRCh38
- UniGene ID
- Hs.632377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092096.1 |
- |
1 |
915 |
|
BC034611.1 |
- |
2 |
344 |
|
View Full Product Details