Gene Transcripts

Gene Symbol
TPRXL
Entrez Gene ID
348825
Gene Name
tetrapeptide repeat homeobox like
Gene Aliases
TPRX3P
Chromosome Location
Chr. 3: 13937307 - 14065984 on Build GRCh38
UniGene ID
Hs.529180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002223.3 3 437
GenBank mRNA AK092426.1 - 3 344
BC027729.2 - 3 529
BC117319.1 - 1 70
DA837466.1 - 3 438

View Full Product Details