Gene Transcripts
- Gene Symbol
- TPRXL
- Entrez Gene ID
- 348825
- Gene Name
- tetrapeptide repeat homeobox like
- Gene Aliases
- TPRX3P
- Chromosome Location
- Chr. 3: 13937307 - 14065984 on Build GRCh38
- UniGene ID
- Hs.529180
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002223.3 |
|
3 |
2138 |
|
GenBank mRNA |
AK092426.1 |
- |
3 |
2063 |
|
BC027729.2 |
- |
5 |
2143 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM764893.1 |
- |
1 |
669 |
|
Chr. 3 - 14065191 - 14065879 [+] |
GenBankmRNA |
BM811485.1 |
- |
|
441 |
|
Chr. - - [-] |
GenBankmRNA |
BX111860.1 |
- |
1 |
487 |
|
Chr. 3 - 14065373 - 14065986 [+] |
View Full Product Details