Gene Transcripts
- Gene Symbol
- TPRXL
- Entrez Gene ID
- 348825
- Gene Name
- tetrapeptide repeat homeobox like
- Gene Aliases
- TPRX3P
- Chromosome Location
- Chr. 3: 13937307 - 14065984 on Build GRCh38
- UniGene ID
- Hs.529180
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002223.3 |
|
3 |
1317 |
|
GenBank mRNA |
AK092426.1 |
- |
3 |
1242 |
|
BC027729.2 |
- |
5 |
1322 |
|
View Full Product Details