Gene Transcripts
- Gene Symbol
- TSG1
- Entrez Gene ID
- 643432
- Gene Name
- tumor suppressor TSG1
- Gene Aliases
- Chromosome Location
- Chr. 6: 93707083 - 93776481 on Build GRCh38
- UniGene ID
- Hs.509936
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015362.1 |
|
8 |
3564 |
|
GenBank mRNA |
AY314745.1 |
- |
8 |
3564 |
|
AY314746.1 |
- |
7 |
3474 |
|
View Full Product Details