Gene Transcripts
- Gene Symbol
- ESRG
- Entrez Gene ID
- 790952
- Gene Name
- embryonic stem cell related (non-protein coding)
- Gene Aliases
- HESRG
- Chromosome Location
- Chr. 3: 54632124 - 54639857 on Build GRCh38
- UniGene ID
- Hs.720658
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027122.1 |
|
4 |
2813 |
|
GenBank mRNA |
DQ445779.2 |
- |
4 |
2812 |
|
KM363598.1 |
- |
4 |
2508 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN429802.1 |
- |
3 |
317 |
|
Chr. 3 - 54632129 - 54634522 [-] |
GENCODE |
ENST00000583516.1 |
- |
4 |
2813 |
|
Chr. 3 - 54632122 - 54639857 [-] |
GenBankmRNA |
BX114820.1 |
- |
2 |
212 |
|
Chr. 3 - 54632293 - 54633716 [-] |
GenBankmRNA |
CN332966.1 |
- |
3 |
292 |
|
Chr. 3 - 54632122 - 54638641 [-] |
View Full Product Details