Gene Transcripts

Gene Symbol
SNHG10
Entrez Gene ID
283596
Gene Name
small nucleolar RNA host gene 10
Gene Aliases
C14orf62, LINC00063, NCRNA00063
Chromosome Location
Chr. 14: 95532912 - 95534872 on Build GRCh38
UniGene ID
Hs.448753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001459.2 2 1212
NR_003138.3 1 1843
GenBank mRNA AK097876.1 - 1 1705
BC036576.2 - 1 1598

View Full Product Details