Gene Transcripts

Gene Symbol
LINC00477
Entrez Gene ID
144360
Gene Name
long intergenic non-protein coding RNA 477
Gene Aliases
C12orf67, FAM191B
Chromosome Location
Chr. 12: 24566964 - 24584168 on Build GRCh38
UniGene ID
Hs.350668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029451.2 1 280
GenBank mRNA AK057456.1 - 1 280
BC104436.1 - 1 260
BC104437.1 - 1 260

View Full Product Details