Gene Transcripts

Gene Symbol
GOLGA8G
Entrez Gene ID
283768
Gene Name
golgin A8 family member G
Gene Aliases
Chromosome Location
Chr. 15: 28519611 - 28532997 on Build GRCh38
UniGene ID
Hs.169639
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033353.1 18 5072
GenBank mRNA AL137398.1 - 1 1703
BC066774.1 - 18 5046
BC099913.1 - 17 5887 4838463

View Full Product Details