Gene Transcripts
- Gene Symbol
- PIN1P1
- Entrez Gene ID
- 5301
- Gene Name
- peptidylprolyl cis/trans isomerase, NIMA-interacting 1 pseudogene 1
- Gene Aliases
- PIN1L
- Chromosome Location
- Chr. 1: 69919322 - 69920317 on Build GRCh38
- UniGene ID
- Hs.654711
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023916.1 |
|
1 |
23 |
|
GenBank mRNA |
AK314840.1 |
- |
1 |
15 |
|
U82382.1 |
- |
1 |
23 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB071534.1 |
- |
1 |
15 |
|
Chr. 1 - 69919330 - 69919644 [+] |
View Full Product Details