Gene Transcripts

Gene Symbol
PIN1P1
Entrez Gene ID
5301
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1 pseudogene 1
Gene Aliases
PIN1L
Chromosome Location
Chr. 1: 69919322 - 69920317 on Build GRCh38
UniGene ID
Hs.654711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_023916.1 1 100
GenBank mRNA AK314840.1 - 1 92
U82382.1 - 1 100


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB071534.1 - 1 92 Chr. 1 - 69919330 - 69919644 [+]
GenBankmRNA CR748481.1 - 1 53 Chr. 1 - 69919369 - 69920015 [+]

View Full Product Details