Gene Transcripts
- Gene Symbol
- FBXL19-AS1
- Entrez Gene ID
- 283932
- Gene Name
- FBXL19 antisense RNA 1 (head to head)
- Gene Aliases
- NCRNA00095
- Chromosome Location
- Chr. 16: 30919319 - 30923269 on Build GRCh38
- UniGene ID
- Hs.711535
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024348.1 |
|
1 |
2725 |
|
GenBank mRNA |
AK090544.1 |
- |
1 |
1639 |
|
AK092263.1 |
- |
1 |
2725 |
|
View Full Product Details