Gene Transcripts

Gene Symbol
TMEM30C
Entrez Gene ID
644444
Gene Name
transmembrane protein 30C
Gene Aliases
CDC50C
Chromosome Location
Chr. 3: 100185824 - 100194186 on Build GRCh38
UniGene ID
Hs.652353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028357.1 1, 2 57
NR_028358.1 1, 2 57
GenBank mRNA AB249666.1 - 1, 2 57
AB250297.1 - 1, 2 57

View Full Product Details