Gene Transcripts

Gene Symbol
HMGB3P1
Entrez Gene ID
128872
Gene Name
high mobility group box 3 pseudogene 1
Gene Aliases
HMG4L, HMGB3L1, dJ18C9.3
Chromosome Location
Chr. 20: 34833575 - 34834462 on Build GRCh38
UniGene ID
Hs.558624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002165.1 1 486
GenBank mRNA BC103849.1 - 1 486
BC103850.1 - 1 485
BC103851.1 - 1 485

View Full Product Details