Gene Transcripts
- Gene Symbol
- SNORA5B
- Entrez Gene ID
- 677795
- Gene Name
- small nucleolar RNA, H/ACA box 5B
- Gene Aliases
- ACA5b
- Chromosome Location
- Chr. 7: 45105968 - 45106099 on Build GRCh38
- UniGene ID
- Hs.693124
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002990.1 |
|
1 |
13 |
|
- Gene Symbol
- TBRG4
- Entrez Gene ID
- 9238
- Gene Name
- transforming growth factor beta regulator 4
- Gene Aliases
- CPR2, FASTKD4
- Chromosome Location
- Chr. 7: 45100100 - 45111747 on Build GRCh38
- UniGene ID
- Hs.231411
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092132.1 |
- |
1 |
1378 |
|
View Full Product Details