Gene Transcripts
- Gene Symbol
- SNORA14A
- Entrez Gene ID
- 677801
- Gene Name
- small nucleolar RNA, H/ACA box 14A
- Gene Aliases
- ACA14a
- Chromosome Location
- Chr. 7: 75943783 - 75943916 on Build GRCh38
- UniGene ID
- Hs.689708
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002955.1 |
|
1 |
79 |
|
GenBank mRNA |
AJ609456.1 |
- |
1 |
79 |
|
View Full Product Details