Gene Transcripts
- Gene Symbol
- CDIPT-AS1
- Entrez Gene ID
- 440356
- Gene Name
- CDIPT antisense RNA 1 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 16: 29863683 - 29868053 on Build GRCh38
- UniGene ID
- Hs.448823
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI825161.1 |
- |
5, 6 |
553 |
|
Chr. 16 - 29863860 - 29868048 [+] |
GenBankmRNA |
BM909113.1 |
- |
|
546 |
|
Chr. - - [-] |
View Full Product Details